Learn More
Fisherbrand™ Alphanumeric Blood Band
Description
- Unique blood bank identification number (BBID) ensures the right specimen goes to the intended patient, minimizing critical transcription errors
- Blood band featuring slim design, quality materials, and smudge-proof labels
- The smudge-proof material protects the patient information for up to 3 days during normal use
- Easily refasten, relocate, and resize with the optional R3™ Reattachment System
Specifications
Specifications
| Closure Type | Snap |
| Material | PET |
| Color | Red |
| Product Type | Alplanumeric Blood Band |