missing translation for 'onlineSavingsMsg'
Learn More

Alphacel Non-Nutritive Bulk, MP Biomedicals

Composed of finely ground cellulose for addition to animal diets to supply bulk with minimum nutritional value

$110.58 - $110.58

Chemical Identifiers

CAS 9004-34-6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN90045305
View Documents
MP Biomedicals Inc
0290045305
5 lb. N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9004-34-6

Specifications

9004-34-6
5 lb.
Alphacel Non-Nutritive Bulk
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'