missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl fluoride, 99%

Catalog No. p-7026241
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2214603 1 g
AAB2214606 5 g
AAB2214614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2214603 Supplier Thermo Scientific Chemicals Supplier No. B2214603
Only null left

CAS: 329-98-6 | C7H7FO2S | 174.189 g/mol

α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

Solubility
Soluble in ether, benzene, anhydrous ethanol, methanol, 2- propane. Hydrolyses in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, amines, bases, water/ moisture. Store at 4°C

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
UN Number UN2923
Beilstein 2088311
Sensitivity Moisture sensitive
Solubility Information Soluble in ether,benzene,anhydrous ethanol,methanol,2- propane. Hydrolyses in water.
Formula Weight 174.19
Percent Purity 99%
Chemical Name or Material alpha-Toluenesulfonyl fluoride
Hazard Category H301-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 206-350-2
RTECSNumber XT8040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only