missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl chloride, 99%

Catalog No. p-7080546
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1012406 5 g
AAA1012414 25 g
AAA1012422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1012406 Supplier Thermo Scientific Chemicals Supplier No. A1012406
Only null left

CAS: 1939-99-7 | C7H7ClO2S | 190.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1939-99-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00007455
InChI Key OAHKWDDSKCRNFE-UHFFFAOYSA-N
Synonym alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
PubChem CID 74740
IUPAC Name phenylmethanesulfonyl chloride
SMILES C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 89°C to 94°C
Boiling Point 120°C (10 mmHg)
Quantity 5 g
UN Number UN3096
Beilstein 972806
Sensitivity Moisture sensitive
Formula Weight 190.65
Percent Purity 99%
Chemical Name or Material alpha-Toluenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 217-717-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only