missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-alpha-Tocopherol, 95%, synthetic

Catalog No. p-4089596
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428120250 25 g Glass bottle
AC428121000 100 g Glass bottle
AC428125000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC428120250 Supplier Thermo Scientific Chemicals Supplier No. 428120250
Only null left

CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 2.5°C to 3.5°C
Density 0.9500g/mL
Boiling Point 393.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 0.95
Merck Index 15, 9653
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in oils, fats, acetone, alcohol,, chloroform, diethyl ether, other fat solvents.
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Viscosity 4200 mPa.s (20°C)
Formula Weight 430.71
Percent Purity 95%
Chemical Name or Material (±)-α-Tocopherol, Synthetic
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 233-466-

RUO – Research Use Only