missing translation for 'onlineSavingsMsg'
Learn More

alpha-Sexithiophene (purified by sublimation), TCI America™

Catalog No. S0504100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0504100MG 100 mg
S05041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0504100MG Supplier TCI America Supplier No. S0504100MG

Chemical Identifiers

CAS 88493-55-4
Molecular Formula C24H14S6
Molecular Weight (g/mol) 494.736
InChI Key KUJYDIFFRDAYDH-UHFFFAOYSA-N
Synonym 6T
PubChem CID 11340899
IUPAC Name 2-thiophen-2-yl-5-[5-[5-(5-thiophen-2-ylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene
SMILES C1=CSC(=C1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)C5=CC=C(S5)C6=CC=CS6

Specifications

Melting Point 304°C
Quantity 100 mg
Formula Weight 494.74
Physical Form Crystalline Powder
Chemical Name or Material alpha-Sexithiophene (purified by sublimation)