missing translation for 'onlineSavingsMsg'
Learn More

alpha-Septithiophene, TCI America™

Catalog No. S0505100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0505100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0505100MG Supplier TCI America Supplier No. S0505100MG

Chemical Identifiers

CAS 86100-63-2
Molecular Formula C28H16S7
Molecular Weight (g/mol) 576.86
MDL Number MFCD09038548
InChI Key DCIYLINCGGGNLC-UHFFFAOYSA-N
PubChem CID 13113982
IUPAC Name 5-(5'-{[2,2'-bithiophen]-5-yl}-[2,2'-bithiophen]-5-yl)-5'-(thiophen-2-yl)-2,2'-bithiophene
SMILES S1C=CC=C1C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Melting Point 328°C
Quantity 100 mg
Formula Weight 576.86
Physical Form Crystalline Powder
Chemical Name or Material alpha-Septithiophene