Learn More
Alpha Scientific™ DIFF-SAFE™ Blood Dispenser for Blood Smears
Catalog No. 22026396
Change view
Click to view available options
Quantity:
100 Pack
25000 Case
Case of 10000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22026396 Case of 10000
22029115 100 Pack
22026397 25000 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22026396 Supplier Alpha Scientific™ Supplier No. DS 101
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cuts the time of smear-making in half, reducing technician fatigue
Cuts the time of smear-making in half, reducing technician fatigue
  • Compared to the typical procedure involving 15 or more steps when sticks or capillaries are used, the procedure using the DIFF-SAFE dispenser cuts the number of steps by two thirds
  • Blunt cannula size is 21G x 0.5 in.
Specifications
Product Type Blood Dispenser
Includes 21G x 0.5 in. Blunt needle
For Use With (Application) Blood smears
Quantity Case of 10000