missing translation for 'onlineSavingsMsg'
Learn More

alpha-Quinquethiophene, TCI America™

Catalog No. Q0079100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0079100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0079100MG Supplier TCI America Supplier No. Q0079100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5660-45-7
Molecular Formula C20H12S5
Molecular Weight (g/mol) 412.616
InChI Key YFBLUJZFRBFQMR-UHFFFAOYSA-N
PubChem CID 395508
IUPAC Name 2,5-bis(5-thiophen-2-ylthiophen-2-yl)thiophene
SMILES C1=CSC(=C1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)C5=CC=CS5

Specifications

Melting Point 253°C
Quantity 100 mg
Formula Weight 412.62
Physical Form Crystalline Powder
Chemical Name or Material alpha-Quinquethiophene