missing translation for 'onlineSavingsMsg'
Learn More

alpha-Pyrone 98.0+%, TCI America™

Catalog No. P11831G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11831G Supplier TCI America Supplier No. P11831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 504-31-4
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.085
MDL Number MFCD00006639
InChI Key ZPSJGADGUYYRKE-UHFFFAOYSA-N
Synonym 2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
PubChem CID 68154
ChEBI CHEBI:37965
IUPAC Name pyran-2-one
SMILES C1=CC(=O)OC=C1

Specifications

Boiling Point 207°C
Quantity 1 g
Formula Weight 96.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Pyrone