missing translation for 'onlineSavingsMsg'
Learn More

(+)-alpha-Pinene, 98%

Catalog No. p-7044163
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0494118 50 g
AAL0494130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0494118 Supplier Thermo Scientific Chemicals Supplier No. L0494118

CAS: 7785-70-8 | C10H16 | 136.24 g/mol

(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

Solubility
Soluble in Ether, Alcohols, Chloroform. Not miscible in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 7785-70-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001346
InChI Key GRWFGVWFFZKLTI-RKDXNWHRSA-N
Synonym +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene
PubChem CID 82227
ChEBI CHEBI:28261
IUPAC Name (1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CC[C@@H]2C[C@H]1C2(C)C

Specifications

Melting Point -62°C
Density 0.858
Boiling Point 154°C to 156°C
Flash Point 32°C (89°F)
Refractive Index 1.466
Quantity 50 g
UN Number UN2368
Beilstein 3648264
Merck Index 14,7445
Solubility Information Soluble in Ether,Alcohols,Chloroform. Not miscible in water.
Formula Weight 136.24
Percent Purity 98%
Chemical Name or Material (+)-alpha-Pinene
Show More Show Less
Hazard Category H226-H304-H315-H317-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H317
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALPHA-PINENE
EINECSNumber 232-087-8
RTECSNumber DT7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only