missing translation for 'onlineSavingsMsg'
Learn More

alpha-Phenylcinnamic Acid 98.0+%, TCI America™

Catalog No. P130025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P130025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P130025G Supplier TCI America Supplier No. P130025G
Only null left

Chemical Identifiers

CAS 91-48-5
Molecular Formula C15H11O2
Molecular Weight (g/mol) 223.25
MDL Number MFCD00004252
InChI Key BIDDLDNGQCUOJQ-SDNWHVSQSA-M
Synonym alpha-phenylcinnamic acid, cis-alpha-phenylcinnamic acid, unii-0pkh62904b, e-2,3-diphenylpropenoic acid, alpha-phenylcinnamic acid, e, 2,3-diphenylacrylic acid, .alpha.-phenylcinnamic acid, .alpha.-stilbenecarboxylic acid, e-2,3-diphenylacrylic acid, trans-2,3-diphenylacrylic acid
PubChem CID 700620
IUPAC Name (2E)-2,3-diphenylprop-2-enoate
SMILES [O-]C(=O)C(=C\C1=CC=CC=C1)\C1=CC=CC=C1

Specifications

Melting Point 175°C
Color White
Quantity 25 g
Formula Weight 224.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Phenylcinnamic Acid