missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphthoflavone 98.0+%, TCI America™
Supplier: TCI America B00561G
Specifications
| alpha-Naphthoflavone | |
| 156°C | |
| C19H12O2 | |
| 1 g | |
| VFMMPHCGEFXGIP-UHFFFAOYSA-N | |
| 2-phenylbenzo[h]chromen-4-one | |
| 11790 | |
| 272.30 | |
| Crystalline Powder |
| 604-59-1 | |
| White-Yellow | |
| MFCD00004985 | |
| alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3 | |
| 272.303 | |
| CHEBI:76995 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 604-59-1 | |
| 272.303 | |
| VFMMPHCGEFXGIP-UHFFFAOYSA-N | |
| 11790 | |
| 2-phenylbenzo[h]chromen-4-one |
| C19H12O2 | |
| MFCD00004985 | |
| alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one | |
| CHEBI:76995 | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3 |
Safety and Handling
RTECSNumber : QL6250000
TSCA : Yes