missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphthoflavone 98.0+%, TCI America™

Catalog No. B00565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00565G 5 g
B00561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00565G Supplier TCI America Supplier No. B00565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 604-59-1
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.303
MDL Number MFCD00004985
InChI Key VFMMPHCGEFXGIP-UHFFFAOYSA-N
Synonym alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one
PubChem CID 11790
ChEBI CHEBI:76995
IUPAC Name 2-phenylbenzo[h]chromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 272.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Naphthoflavone
RTECSNumber QL6250000
TSCA Yes