missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphthoflavone, 97%
CAS: 604-59-1 | C19H12O2 | 272.29 g/mol
$73.25 - $268.66
Chemical Identifiers
| CAS | 604-59-1 |
|---|---|
| Molecular Formula | C19H12O2 |
| Molecular Weight (g/mol) | 272.29 |
| MDL Number | MFCD00004985 |
| InChI Key | VFMMPHCGEFXGIP-UHFFFAOYSA-N |
| Synonym | alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one |
| PubChem CID | 11790 |
| ChEBI | CHEBI:76995 |
| IUPAC Name | 2-phenylbenzo[h]chromen-4-one |
| SMILES | C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC128160010
|
Thermo Scientific Chemicals
128160010 |
1 g | Glass bottle |
Each for $73.25
|
|
||||
|
AC128160050
|
Thermo Scientific Chemicals
128160050 |
5 g | Glass bottle |
Each for $268.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 604-59-1 | |
| 272.29 | |
| VFMMPHCGEFXGIP-UHFFFAOYSA-N | |
| 11790 | |
| 2-phenylbenzo[h]chromen-4-one |
| C19H12O2 | |
| MFCD00004985 | |
| alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one | |
| CHEBI:76995 | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3 |
Specifications
| 604-59-1 | |
| 100.0 | |
| White to Yellow | |
| 99.5% min. (HPLC) | |
| C19H12O2 | |
| 1 g | |
| alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one | |
| VFMMPHCGEFXGIP-UHFFFAOYSA-N | |
| 2-phenylbenzo[h]chromen-4-one | |
| 11790 | |
| 272.29 | |
| Crystalline Powder or Crystals |
| 99.5 | |
| 155°C to 158°C | |
| Authentic | |
| Glass bottle | |
| MFCD00004985 | |
| 17, IV, 5551 | |
| Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3 | |
| 272.29 | |
| CHEBI:76995 | |
| 97% | |
| α-Naphthoflavone, 99.5+% |
Safety and Handling
EINECSNumber : 210-071-1
RTECSNumber : QL6250000
TSCA : TSCA
RUO – Research Use Only