missing translation for 'onlineSavingsMsg'
Learn More

alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%

CAS: 827-36-1 | C10H12N2 | 160.22 g/mol

$56.85 - $56.85

Chemical Identifiers

CAS 827-36-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00013810
InChI Key PAGHXXKYFBGJEH-UHFFFAOYNA-N
Synonym 2-dimethylamino-2-phenylacetonitrile, alpha-dimethylamino phenylacetonitrile, a-dimethylamino phenylacetonitrile, acetonitrile, dimethylamino phenyl, acetonitrile, 2-dimethylamino-2-phenyl, alpha-dimethylaminophenyl acetonitrile, alpha-dimethylaminophenylacetonitrile, a-dimethylaminophenylacetonitrile, dimethylamino phenyl acetonitrile, 2-dimethylamino-2-phenylethanenitrile
PubChem CID 13227
IUPAC Name 2-(dimethylamino)-2-phenylacetonitrile
SMILES CN(C)C(C#N)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2763106
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H2763106
5 g
Each for $56.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

827-36-1
160.22
PAGHXXKYFBGJEH-UHFFFAOYNA-N
13227
CN(C)C(C#N)C1=CC=CC=C1
C10H12N2
MFCD00013810
2-dimethylamino-2-phenylacetonitrile, alpha-dimethylamino phenylacetonitrile, a-dimethylamino phenylacetonitrile, acetonitrile, dimethylamino phenyl, acetonitrile, 2-dimethylamino-2-phenyl, alpha-dimethylaminophenyl acetonitrile, alpha-dimethylaminophenylacetonitrile, a-dimethylaminophenylacetonitrile, dimethylamino phenyl acetonitrile, 2-dimethylamino-2-phenylethanenitrile
2-(dimethylamino)-2-phenylacetonitrile

Specifications

827-36-1
88°C to 90°C (2 mmHg)
C10H12N2
MFCD00013810
UN3276
PAGHXXKYFBGJEH-UHFFFAOYNA-N
2-(dimethylamino)-2-phenylacetonitrile
13227
97%
0.997
104°C (219°F)
1.514
5 g
2-dimethylamino-2-phenylacetonitrile, alpha-dimethylamino phenylacetonitrile, a-dimethylamino phenylacetonitrile, acetonitrile, dimethylamino phenyl, acetonitrile, 2-dimethylamino-2-phenyl, alpha-dimethylaminophenyl acetonitrile, alpha-dimethylaminophenylacetonitrile, a-dimethylaminophenylacetonitrile, dimethylamino phenyl acetonitrile, 2-dimethylamino-2-phenylethanenitrile
CN(C)C(C#N)C1=CC=CC=C1
160.22
160.22
alpha-(N,N-Dimethylamino)phenylacetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : AL9625000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only