missing translation for 'onlineSavingsMsg'
Learn More

alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%

Catalog No. AAH2763106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2763106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2763106 Supplier Thermo Scientific Chemicals Supplier No. H2763106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827-36-1 | C10H12N2 | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 827-36-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00013810
InChI Key PAGHXXKYFBGJEH-UHFFFAOYNA-N
Synonym 2-dimethylamino-2-phenylacetonitrile, alpha-dimethylamino phenylacetonitrile, a-dimethylamino phenylacetonitrile, acetonitrile, dimethylamino phenyl, acetonitrile, 2-dimethylamino-2-phenyl, alpha-dimethylaminophenyl acetonitrile, alpha-dimethylaminophenylacetonitrile, a-dimethylaminophenylacetonitrile, dimethylamino phenyl acetonitrile, 2-dimethylamino-2-phenylethanenitrile
PubChem CID 13227
IUPAC Name 2-(dimethylamino)-2-phenylacetonitrile
SMILES CN(C)C(C#N)C1=CC=CC=C1

Specifications

Density 0.997
Boiling Point 88°C to 90°C (2 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.514
Quantity 5 g
UN Number UN3276
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material alpha-(N,N-Dimethylamino)phenylacetonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
RTECSNumber AL9625000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only