missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylphenylacetonitrile, 96%

Catalog No. AAA1363614
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363614 25 g
AAA1363609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1363614 Supplier Thermo Scientific Chemicals Supplier No. A1363614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1823-91-2 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1823-91-2
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001871
InChI Key NVAOLENBKNECGF-UHFFFAOYNA-N
Synonym 2-phenylpropionitrile, hydratroponitrile, alpha-methylphenylacetonitrile, alpha-methylbenzyl cyanide, 2-phenylpropiononitrile, benzeneacetonitrile, .alpha.-methyl, methylbenzyl cyanide, .alpha.-phenylpropionitrile, .alpha.-phenylethyl cyanide, .alpha.-methylbenzyl cyanide
PubChem CID 15761
IUPAC Name 2-phenylpropanenitrile
SMILES CC(C#N)C1=CC=CC=C1

Specifications

Density 0.985
Boiling Point 230°C to 232°C
Flash Point 93°C (199°F)
Refractive Index 1.5106
Quantity 25 g
UN Number UN3276
Beilstein 774576
Formula Weight 131.18
Percent Purity 97%
Chemical Name or Material alpha-Methylphenylacetonitrile
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H331-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 217-354-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only