missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyllevulinic Acid 98.0+%, TCI America™

Catalog No. m10585g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10585G Supplier TCI America Supplier No. M10585G
Only null left

Chemical Identifiers

CAS 6641-83-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00143584
InChI Key UZTJTTKEYGHTNM-UHFFFAOYSA-N
Synonym 2-Methyl-4-oxopentanoic Acid
PubChem CID 225902
IUPAC Name 2-methyl-4-oxopentanoic acid
SMILES CC(CC(=O)C)C(=O)O

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Methyllevulinic Acid