missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylhydrocinnamic acid, 98%

Catalog No. p-3212360
missing translation for 'changeView'
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379080010 1 g Glass bottle
AC379080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC379080010 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' 379080010
Only null left

CAS: 1009-67-2 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1009-67-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00192301
InChI Key MCIIDRLDHRQKPH-UHFFFAOYNA-N
Synonym 2-benzylpropionic acid, alpha-methylhydrocinnamic acid, 2-methyl-3-phenylpropionic acid, 2-benzylpropanoic acid, 2-methyl-3-phenyl-propionic acid, acmc-20ao3h, alpha-methylhydrocinnamicacid, alpha-methyl-hydrocinnamic acid, 2-methyl-3-phenyl-propanoic acid, #
PubChem CID 99862
IUPAC Name 2-methyl-3-phenylpropanoic acid
SMILES CC(CC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color White to Yellow
Boiling Point 167°C to 168°C (23.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(CH3)CO2H
Quantity 1 g
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Methylhydrocinnamic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only