missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-valerolactone (stabilized with HQ) 97.0+%, TCI America™

Catalog No. M14535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14535G 5 g
M145325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14535G Supplier TCI America Supplier No. M14535G
Only null left

Chemical Identifiers

CAS 62873-16-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD06797118
InChI Key KYLUHLJIAMFYKW-UHFFFAOYNA-N
Synonym 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone
PubChem CID 99939
IUPAC Name 5-methyl-3-methylideneoxolan-2-one
SMILES CC1CC(=C)C(=O)O1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Methylene-gamma-valerolactone (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator