missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamic Acid 98.0+%, TCI America™

Catalog No. p-7132670
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13365G 5 g
M133625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13365G Supplier TCI America Supplier No. M13365G
Only null left

Chemical Identifiers

CAS 1199-77-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002652
InChI Key XNCRUNXWPDJHGV-BQYQJAHWSA-N
Synonym alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid
PubChem CID 637817
IUPAC Name (2E)-2-methyl-3-phenylprop-2-enoic acid
SMILES C\C(=C/C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 288°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Methylcinnamic Acid