Learn More
alpha-Methylcinnamic acid, 99%
CAS: 1199-77-5 | C10H10O2 | 162.19 g/mol
Supplier: Thermo Scientific Chemicals L0080606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| alpha-Methylcinnamic acid | |
| 78°C to 82°C | |
| C10H10O2 | |
| 5 g | |
| alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid | |
| C\C(=C/C1=CC=CC=C1)C(O)=O | |
| 162.19 | |
| 162.19 |
| 1199-77-5 | |
| 288°C | |
| MFCD00002652 | |
| 2042544 | |
| XNCRUNXWPDJHGV-BQYQJAHWSA-N | |
| (E)-2-methyl-3-phenylprop-2-enoic acid | |
| 637817 | |
| 99% |
Chemical Identifiers
| 1199-77-5 | |
| 162.19 | |
| XNCRUNXWPDJHGV-BQYQJAHWSA-N | |
| 637817 | |
| C\C(=C/C1=CC=CC=C1)C(O)=O |
| C10H10O2 | |
| MFCD00002652 | |
| alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid | |
| (E)-2-methyl-3-phenylprop-2-enoic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-847-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only