missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Methyl-L-proline 96.0+%, TCI America™
$516.35 - $1787.63
Chemical Identifiers
| CAS | 42856-71-3 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.159 |
| MDL Number | MFCD01318647 |
| InChI Key | LWHHAVWYGIBIEU-LURJTMIESA-N |
| PubChem CID | 6993664 |
| IUPAC Name | (2S)-2-methylpyrrolidine-2-carboxylic acid |
| SMILES | CC1(CCCN1)C(=O)O |
Chemical Identifiers
| 42856-71-3 | |
| 129.159 | |
| LWHHAVWYGIBIEU-LURJTMIESA-N | |
| (2S)-2-methylpyrrolidine-2-carboxylic acid |
| C6H11NO2 | |
| MFCD01318647 | |
| 6993664 | |
| CC1(CCCN1)C(=O)O |
Specifications
| 42856-71-3 | |
| White | |
| MFCD01318647 | |
| LWHHAVWYGIBIEU-LURJTMIESA-N | |
| (2S)-2-methylpyrrolidine-2-carboxylic acid | |
| 6993664 | |
| ≥96.0% (T) | |
| alpha-Methyl-L-proline |
| 310°C | |
| C6H11NO2 | |
| 1 g | |
| CC1(CCCN1)C(=O)O | |
| 129.159 | |
| 129.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No