missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-gamma-butyrolactone 98.0+%, TCI America™

Catalog No. M110025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M110025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M110025G Supplier TCI America Supplier No. M110025G

Chemical Identifiers

CAS 1679-47-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00005396
InChI Key QGLBZNZGBLRJGS-UHFFFAOYNA-N
Synonym Dihydro-3-methyl-2(3H)-furanone
PubChem CID 98323
IUPAC Name 3-methyloxolan-2-one
SMILES CC1CCOC1=O

Specifications

Physical Form Clear Liquid at 20°C
Boiling Point 80°C
Quantity 25 g
Color Colorless
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Chemical Name or Material alpha-Methyl-gamma-butyrolactone