missing translation for 'onlineSavingsMsg'
Learn More

alpha-Mangostin 97.0+%, TCI America™

Catalog No. p-7133433
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M279310MG 10 mg
M2793100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M279310MG Supplier TCI America Supplier No. M279310MG

Chemical Identifiers

CAS 6147-11-1
Molecular Formula C24H26O6
Molecular Weight (g/mol) 410.47
MDL Number MFCD00135200
InChI Key GNRIZKKCNOBBMO-UHFFFAOYSA-N
PubChem CID 5281650
ChEBI CHEBI:67547
IUPAC Name 1,3,6-trihydroxy-7-methoxy-2,8-bis(3-methylbut-2-enyl)xanthen-9-one
SMILES CC(=CCC1=C(C=C2C(=C1O)C(=O)C3=C(C(=C(C=C3O2)O)OC)CC=C(C)C)O)C

Specifications

Melting Point 180°C
Color Yellow
Quantity 10 mg
Formula Weight 410.47
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Mangostin
RTECSNumber ZD6122420
TSCA No
Recommended Storage Refrigerator