missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ionone 90.0+%, TCI America™

Catalog No. p-7131198
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I007625G 25 g
I0076250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I007625G Supplier TCI America Supplier No. I007625G
Only null left

Chemical Identifiers

CAS 127-41-3
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.30
MDL Number MFCD00001565
InChI Key UZFLPKAIBPNNCA-BQYQJAHWNA-N
Synonym alpha-ionone, iraldeine, e-alpha-ionone, irisone, alpha-ionon, trans-alpha-ionone, alpha-e-ionone, alpha-ionone natural, beta-lonone, ionone, alpha
PubChem CID 5282108
ChEBI CHEBI:32319
IUPAC Name (3E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
SMILES CC(=O)\C=C\C1C(C)=CCCC1(C)C

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 192.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Ionone
EINECSNumber (3)-2387
RTECSNumber EN0525000
TSCA Yes