missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ionone, 90+%

Catalog No. AAA1709822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1709822 100 g
AAA1709836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1709822 Supplier Thermo Scientific Chemicals Supplier No. A1709822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 127-41-3 | C13H20O | 192.30 g/mol

alpha-Ionone, is used in many floral accords. Gives a violet flower nuance to compositions, but is also used in rose, woody, amber, balsamic, and powdery fragrances. It is also used in perfumery, flavoring, and vitamin A (retinol) production for cosmetics and toiletries. beta isomer shows a less floral odor than the alpha. It has good substantivity and is used in all areas of perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Ionone, is used in many floral accords. Gives a violet flower nuance to compositions, but is also used in rose, woody, amber, balsamic, and powdery fragrances. It is also used in perfumery, flavoring, and vitamin A (retinol) production for cosmetics and toiletries. beta isomer shows a less floral odor than the alpha. It has good substantivity and is used in all areas of perfumery.

Solubility
Soluble in ethanol fixed oils, propylene glycol. Slightly soluble in alcohol, ether, mineral oil.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 127-41-3
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.30
MDL Number MFCD00001565
InChI Key UZFLPKAIBPNNCA-BQYQJAHWNA-N
Synonym alpha-ionone, iraldeine, e-alpha-ionone, irisone, alpha-ionon, trans-alpha-ionone, alpha-e-ionone, alpha-ionone natural, beta-lonone, ionone, alpha
PubChem CID 5282108
ChEBI CHEBI:32319
IUPAC Name (E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
SMILES CC(=O)\C=C\C1C(C)=CCCC1(C)C

Specifications

Density 0.931
Boiling Point 237°C
Flash Point 117°C (243°F)
Refractive Index 1.498
Quantity 100 g
Beilstein 2046084
Merck Index 14,5056
Solubility Information Soluble in ethanol fixed oils,propylene glycol. Slightly soluble in alcohol,ether,mineral oil.
Formula Weight 192.3
Percent Purity ≥90%
Chemical Name or Material alpha-Ionone
Show More Show Less
EINECSNumber 204-841-6
RTECSNumber EN0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only