missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hydroxy-gamma-butyrolactone 98.0+%, TCI America™

Catalog No. p-7130722
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H078810G 10 g
H078825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H078810G Supplier TCI America Supplier No. H078810G
Only null left

Chemical Identifiers

CAS 19444-84-9
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00134268,MFCD00211245
InChI Key FWIBCWKHNZBDLS-UHFFFAOYNA-N
Synonym alpha-hydroxy-gamma-butyrolactone, 3-hydroxydihydrofuran-2 3h-one, 2-hydroxy-gamma-butyrolactone, 3-hydroxytetrahydrofuran-2-one, 2 3h-furanone, dihydrohydroxy, 3s-3-hydroxydihydrofuran-2 3h-one, 3-hydroxydihydro-2 3h-furanone, hydroxybutyrolactone, r-dihydro-3-hydroxyfuran-2 3h-one, 2 3h-furanone, dihydro-3-hydroxy
PubChem CID 545831
IUPAC Name 3-hydroxyoxolan-2-one
SMILES OC1CCOC1=O

Specifications

Boiling Point 133°C
Quantity 10 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Hydroxy-gamma-butyrolactone