missing translation for 'onlineSavingsMsg'
Learn More

alpha-Fluorophenylacetic acid, 97%

Catalog No. AAL01745MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL01745MC 100 mg
AAL01745ME 500 mg
AAL0174504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL01745MC Supplier Thermo Scientific Chemicals Supplier No. L01745MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1578-63-8 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1578-63-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004221
InChI Key ATPPNMLQNZHDOG-UHFFFAOYSA-N
Synonym alpha-fluorophenylacetic acid, fluorophenylacetic acid, fluoro phenyl acetic acid, .alpha.-fluorophenylacetic acid, benzeneacetic acid, .alpha.-fluoro, fluorophenyl acetic acid, a-fluorophenylacetic acid, a-fluorobenzeneacetic acid, a-fluorophenyl acetic acid, acmc-1bt6i
PubChem CID 102649
IUPAC Name 2-fluoro-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)F

Specifications

Melting Point 78°C to 82°C
Quantity 100 mg
UN Number UN2811
Beilstein 2087378
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material alpha-Fluorophenylacetic acid
Hazard Category H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 216-420-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only