missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Endorphin, human

Catalog No. AAJ63462MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63462MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63462MCR Supplier Thermo Scientific Chemicals Supplier No. J63462MCR

 

An opioid peptide and substrate for transglutaminase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Common Name β-Lipotropin (61-76)
Format Powder
For Use With (Application) An opioid peptide and substrate for transglutaminase
Molecular Weight (g/mol) 1745.98
Product Type alpha-Endorphin, human
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence H-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-OH

RUO – Research Use Only