missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Galactose pentaacetate

Catalog No. p-7043227
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6636606 5 g
AAJ6636614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6636606 Supplier Thermo Scientific Chemicals Supplier No. J6636606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4163-59-1 | C16H22O11 | 390.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4163-59-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose
PubChem CID 6432426
ChEBI CHEBI:63145
IUPAC Name [(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 143°C to 144°C
Color White
Quantity 5 g
Solubility Information Soluble in methanol at approximately 50mg/ml
Formula Weight 390.34
Physical Form Powder
Chemical Name or Material alpha-D-Galactose pentaacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only