missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclopropylbenzyl Alcohol 96.0+%, TCI America™

Catalog No. p-7124725
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09865ML 5 mL
C098625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09865ML Supplier TCI America Supplier No. C09865ML
Only null left

Chemical Identifiers

CAS 1007-03-0
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00001299
InChI Key GOXKCYOMDINCCD-UHFFFAOYSA-N
Synonym cyclopropyl phenyl methanol, alpha-cyclopropylbenzyl alcohol, cyclopropylphenylcarbinol, cyclopropyl phenylmethanol, cyclopropyl phenyl carbinol, phenylcyclopropyl methanol, acmc-2097rv, cyclopropylphenylmethan-1-ol, alpha-cyclopropylbenzylalcohol, cyclopropyl phenyl methanol #
PubChem CID 66090
IUPAC Name cyclopropyl(phenyl)methanol
SMILES C1CC1C(C2=CC=CC=C2)O

Specifications

Color Colorless
Boiling Point 132°C
Quantity 5 mL
Formula Weight 148.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Cyclopropylbenzyl Alcohol