missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Cyclohexyl-DL-mandelic Acid 98.0+%, TCI America™
$134.75 - $411.98
Chemical Identifiers
| CAS | 4335-77-7 |
|---|---|
| Molecular Formula | C14H18O3 |
| Molecular Weight (g/mol) | 234.295 |
| MDL Number | MFCD00194543 |
| InChI Key | YTRNSQPXEDGWMR-UHFFFAOYSA-N |
| Synonym | alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32 |
| PubChem CID | 97700 |
| IUPAC Name | 2-cyclohexyl-2-hydroxy-2-phenylacetic acid |
| SMILES | C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O |
Chemical Identifiers
| 4335-77-7 | |
| 234.295 | |
| YTRNSQPXEDGWMR-UHFFFAOYSA-N | |
| 97700 | |
| C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O |
| C14H18O3 | |
| MFCD00194543 | |
| alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32 | |
| 2-cyclohexyl-2-hydroxy-2-phenylacetic acid |
Specifications
| 4335-77-7 | |
| White-Yellow | |
| C14H18O3 | |
| 5 g | |
| YTRNSQPXEDGWMR-UHFFFAOYSA-N | |
| 2-cyclohexyl-2-hydroxy-2-phenylacetic acid | |
| 97700 | |
| ≥98.0% (HPLC,T) | |
| alpha-Cyclohexyl-DL-mandelic Acid |
| 167°C | |
| 135°C | |
| MFCD00194543 | |
| alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32 | |
| C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O | |
| 234.295 | |
| 234.30 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1902
RTECSNumber : GV2770000
TSCA : No