missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin 98.0+%, TCI America™

Supplier:  TCI America C077625G

 View more versions of this product

Catalog No. C077625G


Only null left

Description

Description

Specifications

Specifications

alpha-Cyclodextrin
White
MFCD00078207
cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
320760
≥98.0% (HPLC)
10016-20-3
C36H60O30
25 g
HFHDHCJBZVLPGP-UHFFFAOYSA-N
972.846
972.85
Crystalline Powder

Chemical Identifiers

10016-20-3
972.846
HFHDHCJBZVLPGP-UHFFFAOYSA-N
320760
C36H60O30
MFCD00078207
cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0101

RTECSNumber : GU2292000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions