missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin 98.0+%, TCI America™

Catalog No. C077610G
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C077610G 10 g
C077625G 25 g
C0776100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C077610G Supplier TCI America Supplier No. C077610G

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
MDL Number MFCD00078207
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 10 g
Formula Weight 972.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclodextrin
EINECSNumber (8)-0101
RTECSNumber GU2292000
TSCA Yes