missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin, 98%, Spectrum™ Chemical

Catalog No. p-7157186
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600438 500 g Amber Glass Bottle
18600439 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600438 Supplier Spectrum Chemical Mfg Cor Supplier No. C1620500GMBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1620, 10016-20-3, C36H60O30

Spectrum™ Chemical alpha-Cyclodextrin , a polysaccharide, can be used as a dietary fiber. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.85
InChI Key HFHDHCJBZVLPGP-UHFFFAOYNA-N
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Water 10%