missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Caryophyllene 93.0+%, TCI America™
Supplier: TCI America C09571ML
Specifications
| alpha-Caryophyllene | |
| Yellow | |
| C15H24 | |
| 1 mL | |
| FAMPSKZZVDUYOS-HRGUGZIWSA-N | |
| (1E,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene | |
| 123132308 | |
| ≥93.0% (GC) |
| 6753-98-6 | |
| 123°C | |
| MFCD00042689 | |
| alpha-Humulene | |
| C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1 | |
| 204.36 | |
| 204.36 | |
| Liquid |
Chemical Identifiers
| 6753-98-6 | |
| 204.36 | |
| FAMPSKZZVDUYOS-HRGUGZIWSA-N | |
| 123132308 | |
| C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1 |
| C15H24 | |
| MFCD00042689 | |
| alpha-Humulene | |
| (1E,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene |
Safety and Handling
EINECSNumber : (4)-0592
RTECSNumber : GZ4817500
TSCA : Yes