missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alpha-Bungarotoxin Conjugates

Catalog No. T1175
Encompass
Change view
Click to view available options
Label or Dye:
Alexa Fluor™ 488
Alexa Fluor™ 555
Alexa Fluor™ 594
Alexa Fluor™ 647
Biotin-XX
Tetramethylrhodamine
Unlabeled
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
T1175 Tetramethylrhodamine
B13422 Alexa Fluor™ 488
B35451 Alexa Fluor™ 555
B13423 Alexa Fluor™ 594
B35450 Alexa Fluor™ 647
B1196 Biotin-XX
B1601 Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. T1175 Supplier Invitrogen™ Supplier No. T1175
  • ONLINE EXCLUSIVE PRICE
    $844.00
    $692.08 / Each

    Save $151.92

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor.

Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

Specifications

Color Yellow
Detection Method Fluorescence
Product Type Tetramethylrhodamine α-Bungarotoxin
Quantity 500 μg
Content And Storage Store in freezer (–5° to –30°C) and protect from light.
Label or Dye Tetramethylrhodamine
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Receptors
Excitation/Emission 554/577 nm