missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromocinnamaldehyde 98.0+%, TCI America™

Catalog No. B125325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B125325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B125325G Supplier TCI America Supplier No. B125325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5443-49-2
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD00006965
InChI Key WQRWNOKNRHCLHV-TWGQIWQCSA-N
Synonym alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo
PubChem CID 5369403
IUPAC Name (Z)-2-bromo-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=C(C=O)Br

Specifications

Melting Point 70°C
Quantity 25 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Bromocinnamaldehyde
EINECSNumber (3)-1149
RTECSNumber GD6480000
TSCA No