Learn More
alpha-Bromocinnamaldehyde, 98%
CAS: 5443-49-2 | C9H7BrO | 211.058 g/mol
Supplier: Thermo Scientific Chemicals A1423022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| alpha-Bromocinnamaldehyde | |
| 68°C to 72°C | |
| MFCD00006965 | |
| Air Sensitive | |
| WQRWNOKNRHCLHV-TWGQIWQCSA-N | |
| (Z)-2-bromo-3-phenylprop-2-enal | |
| 5369403 | |
| 98% |
| 5443-49-2 | |
| C9H7BrO | |
| 100 g | |
| alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo | |
| C1=CC=C(C=C1)C=C(C=O)Br | |
| 211.058 | |
| 211.06 |
Chemical Identifiers
| 5443-49-2 | |
| 211.058 | |
| WQRWNOKNRHCLHV-TWGQIWQCSA-N | |
| 5369403 | |
| C1=CC=C(C=C1)C=C(C=O)Br |
| C9H7BrO | |
| MFCD00006965 | |
| alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo | |
| (Z)-2-bromo-3-phenylprop-2-enal |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 226-637-6
RTECSNumber : GD6480000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only