missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromo-gamma-butyrolactone 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 140°C |
| Quantity | 25 g |
| Formula Weight | 164.99 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | alpha-Bromo-gamma-butyrolactone |
Safety and Handling
| EINECSNumber | (5)-6756 |
| TSCA | Yes |