missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-4-nitro-o-cresol, 95%, Thermo Scientific™

Chemical Identifiers

CAS 772-33-8
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00007340
InChI Key KFDPCYZHENQOBV-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitrobenzyl bromide, 2-bromomethyl-4-nitrophenol, alpha-bromo-4-nitro-o-cresol, phenol, 2-bromomethyl-4-nitro, koshland reagent i, 2-bromomethyl-4-nitro-phenol, 2-hydroxy-5-nitrobenzylbromide, .alpha.-bromo-4-nitro-o-cresol, o-cresol, .alpha.-bromo-4-nitro, koshland i
PubChem CID 13044
IUPAC Name 2-(bromomethyl)-4-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CBr)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC388520050
SDS View Documents
Thermo Scientific Chemicals
388520050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC388520250
SDS View Documents
Thermo Scientific Chemicals
388520250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

772-33-8
232.033
KFDPCYZHENQOBV-UHFFFAOYSA-N
13044
C1=CC(=C(C=C1[N+](=O)[O-])CBr)O
C7H6BrNO3
MFCD00007340
2-hydroxy-5-nitrobenzyl bromide, 2-bromomethyl-4-nitrophenol, alpha-bromo-4-nitro-o-cresol, phenol, 2-bromomethyl-4-nitro, koshland reagent i, 2-bromomethyl-4-nitro-phenol, 2-hydroxy-5-nitrobenzylbromide, .alpha.-bromo-4-nitro-o-cresol, o-cresol, .alpha.-bromo-4-nitro, koshland i
2-(bromomethyl)-4-nitrophenol

Specifications

772-33-8
100.0
95%
C7H6BrNO3
5g
KFDPCYZHENQOBV-UHFFFAOYSA-N
2-(bromomethyl)-4-nitrophenol
13044
≥94%
94.0
Authentic
Glass bottle
MFCD00007340
2-hydroxy-5-nitrobenzyl bromide, 2-bromomethyl-4-nitrophenol, alpha-bromo-4-nitro-o-cresol, phenol, 2-bromomethyl-4-nitro, koshland reagent i, 2-bromomethyl-4-nitro-phenol, 2-hydroxy-5-nitrobenzylbromide, .alpha.-bromo-4-nitro-o-cresol, o-cresol, .alpha.-bromo-4-nitro, koshland i
C1=CC(=C(C=C1[N+](=O)[O-])CBr)O
232.033
232.03
α-Bromo-4-nitro-o-cresol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only