missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2-chlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. B295025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B295025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B295025G Supplier TCI America Supplier No. B295025G
Only null left

Chemical Identifiers

CAS 29270-30-2
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD03428513
InChI Key XHAPROULWZYBGA-UHFFFAOYSA-N
Synonym alpha-bromo-2-chlorophenylacetic acid, 2-bromo-2-2-chlorophenyl acetic acid, 2-bromo-2-2'-chlorophenyl acetic acid, a-bromo 2-chlorophenylacetic acid, alpha-bromo-2-chlorophenyl acetic acid, alpha-bromo 2-chloro phenylacetic acid, bromo 2-chlorophenyl acetic acid, a-bromo-2-chlorophenylacetic acid, alapha-bromo-2-chloro-phenylacetic acid, benzeneacetic acid, alpha-bromo-2-chloro
PubChem CID 2774031
IUPAC Name 2-bromo-2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)Br)Cl

Specifications

Melting Point 110°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 3261
Formula Weight 249.49
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material alpha-Bromo-2-chlorophenylacetic Acid