missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzoin Oxime 98.0+%, TCI America™

Catalog No. p-7118340
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B008125G 25 g
B0081500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B008125G Supplier TCI America Supplier No. B008125G
Only null left

Chemical Identifiers

CAS 441-38-3
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00004501
InChI Key WAKHLWOJMHVUJC-FYWRMAATNA-N
Synonym 2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
PubChem CID 7057888
IUPAC Name (2E)-2-(N-hydroxyimino)-1,2-diphenylethan-1-ol
SMILES O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C
Color White
Quantity 25 g
Formula Weight 227.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Benzoin Oxime
EINECSNumber (4)-0287
RTECSNumber DI1750000
TSCA Yes