missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzoin oxime, 98+%

Catalog No. p-7080898
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1063014 25 g
AAA1063022 100 g
AAA1063036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1063014 Supplier Thermo Scientific Chemicals Supplier No. A1063014
Only null left

CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 441-38-3
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00004501
InChI Key WAKHLWOJMHVUJC-FYWRMAATNA-N
Synonym 2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
PubChem CID 7057888
IUPAC Name (1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
SMILES O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 151°C to 154°C
Quantity 25 g
Beilstein 2051941
Sensitivity Light sensitive
Merck Index 14,1094
Formula Weight 227.26
Percent Purity ≥98%
Chemical Name or Material alpha-Benzoin oxime
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 207-127-2
RTECSNumber DI1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only