missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzoin oxime, 98%

Catalog No. AC105531000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105531000 100 g Glass bottle
AC105530250 25 g Glass bottle
AC105535000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105531000 Supplier Thermo Scientific Chemicals Supplier No. 105531000
Only null left

CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 441-38-3
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00004501
InChI Key WAKHLWOJMHVUJC-FYWRMAATNA-N
Synonym 2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
PubChem CID 7057888
IUPAC Name (1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
SMILES O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(OH)C(=NOH)C6H5
Quantity 100 g
Beilstein 08,IV,1282
Merck Index 15,1096
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,aq. ammonium hydroxide
Formula Weight 227.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Benzoin oxime
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
EINECSNumber 207-127-2

RUO – Research Use Only