missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzaldoxime 90.0+%, TCI America™

Catalog No. B001125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001125G 25 g
B0011100G 100 g
B0011500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B001125G Supplier TCI America Supplier No. B001125G
Only null left

Chemical Identifiers

CAS 622-31-1
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00002119
InChI Key VTWKXBJHBHYJBI-SOFGYWHQSA-N
Synonym benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime
PubChem CID 5324611
IUPAC Name (NE)-N-benzylidenehydroxylamine
SMILES C1=CC=C(C=C1)C=NO

Specifications

Color White-Yellow
Boiling Point 123°C
Quantity 25 g
Formula Weight 121.14
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material alpha-Benzaldoxime