missing translation for 'onlineSavingsMsg'
Learn More

alpha-Amylcinnamaldehyde 90.0+%, TCI America™

Catalog No. p-7117088
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A085125ML 25 mL
A0851500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A085125ML Supplier TCI America Supplier No. A085125ML
Only null left

Chemical Identifiers

CAS 122-40-7
Molecular Formula C14H18O
Molecular Weight (g/mol) 202.297
MDL Number MFCD00006988
InChI Key HMKKIXGYKWDQSV-KAMYIIQDSA-N
Synonym Jasminal, alpha-Pentylcinnamaldehyde, 2-Amyl-3-phenylacrylaldehyde, 2-Amyl-3-phenyl-2-propenal
PubChem CID 1712058
ChEBI CHEBI:32318
IUPAC Name (2Z)-2-benzylideneheptanal
SMILES CCCCCC(=CC1=CC=CC=C1)C=O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 3082
Formula Weight 202.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Amylcinnamaldehyde
EINECSNumber (3)-2657
RTECSNumber GD6825000
TSCA Yes