missing translation for 'onlineSavingsMsg'
Learn More

alpha-Amino-gamma-butyrolactone Hydrobromide 98.0+%, TCI America™

Catalog No. A02835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02835G 5 g
A02831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A02835G Supplier TCI America Supplier No. A02835G

Chemical Identifiers

CAS 6305-38-0
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.017
MDL Number MFCD00012723
InChI Key MKLNTBLOABOJFZ-UHFFFAOYSA-N
Synonym alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
PubChem CID 73508
IUPAC Name 3-aminooxolan-2-one;hydrobromide
SMILES C1COC(=O)C1N.Br

Specifications

Melting Point 237°C
Color White
Quantity 5 g
Formula Weight 182.02
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Amino-gamma-butyrolactone Hydrobromide